o-Xylene, Spectrophotometric Grade, 96% min
o-Xylene, Spectrophotometric Grade, 96% min
o-Xylene, Spectrophotometric Grade, 96% min
Thermo Scientific Chemicals

o-Xylene, Spectrophotometric Grade, 96% min

CAS: 95-47-6 | C8H10 | 106.17 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 L
4 L
產品號碼 032471.K7
亦稱為 032471-K7
價格 (TWD)
-
Quantity:
4 L
Request bulk or custom format
化學識別
CAS95-47-6
IUPAC Name1,2-xylene
Molecular FormulaC8H10
InChI KeyCTQNGGLPUBDAKN-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C
檢視更多
規格Specification Sheet規格表
Residue on Evaporation5ppm
Residual water0.05%
o-Xylene is used as a solvent for paints and adhesives, and to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only