Hydrochloric acid 1.0 N, Biotech Reagent, GMP, J.T.Baker™
Avantor™
Hydrochloric acid 1.0 N, Biotech Reagent, GMP, J.T.Baker™
Made from USP purified water and NF (Multi-Compendial) hydrochloric acid. Meets BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.Más información
Have Questions?
Cambiar vista
Cantidad:
19 L
200 L
Envase:
Ver todo
Polyethylene drum
Número de catálogo 0325-09
Precio (USD)
-
Solicite un presupuesto
Cantidad:
200 L
Envase:
Polyethylene drum
Made from USP purified water and NF (Multi-Compendial) hydrochloric acid. Meets BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.
Especificaciones
Punto de ebullición623°C (1013 hPa)
CAS7647-01-0
Nombre del producto químico o materialHydrochloric acid