Benzyl azide, 94%, Thermo Scientific Chemicals
Benzyl azide, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyl azide, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 g
10 g
Catalog number 032635.18
also known as 032635-18
Price (USD)/ Each
584.65
Online exclusive
688.00 
Save 103.35 (15%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
584.65
Online exclusive
688.00 
Save 103.35 (15%)
Add to cart
Benzyl azide, 94%, Thermo Scientific Chemicals
Catalog number032635.18
Price (USD)/ Each
584.65
Online exclusive
688.00 
Save 103.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(azidomethyl)benzene
Molecular FormulaC7H7N3
InChI KeyUDLLFLQFQMACJB-UHFFFAOYSA-N
SMILES[N-]=[N+]=NCC1=CC=CC=C1
Molecular Weight (g/mol)133.15
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥92.0%
Refractive Index1.5255-1.5325 @ 20?C
Appearance (Color)Clear colorless to pale brown
FormLiquid
Infrared spectrumConforms
Benzyl azide is used in the synthesis of 1,2,3-triazole derivatives. It serves as a reagent used to introduce a PhCH2N group into a molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only