Bromophenol Blue sodium salt, ACS
Bromophenol Blue sodium salt, ACS
Bromophenol Blue sodium salt, ACS
Thermo Scientific Chemicals

Bromophenol Blue sodium salt, ACS

CAS: 34725-61-6
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
產品號碼 032639.14
亦稱為 032639-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS34725-61-6
IUPAC Namesodium 3,5-dibromo-4-[3-(2,6-dibromo-4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular FormulaC19H9Br4NaO5S
InChI KeyKGANHHDZDYZJEM-UHFFFAOYNA-M
SMILES[Na+].OC1=CC(Br)=C(C(Br)=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=C(Br)C=C([O-])C=C1Br
檢視更多
規格Specification Sheet規格表
CommentVisual transition interval pH 3.0 (yellow) to pH 4.6 (blue)
CommentClarity of solution P.T.
Bromophenol Blue sodium salt is used as a tracking dye for protein and nucleic acid electrophoresis in polyacrylamide gels. It also used in the preparation of protein samples for western blotting analysis. It finds application in staining histones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only