Diclorhidrato de hidracina, > 98 %, Thermo Scientific Chemicals
Diclorhidrato de hidracina, > 98 %, Thermo Scientific Chemicals
Diclorhidrato de hidracina, > 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diclorhidrato de hidracina, > 98 %, Thermo Scientific Chemicals

Para usar como eliminador de cloro en flujos de gas HCl
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
Número de catálogo 032725.22
también denominado 032725-22
Precio (EUR)
76,00
Each
Cantidad:
100 g
Pedido a granel o personalizado
Precio (EUR)
76,00
Each
Identificadores químicos
CAS5341-61-7
IUPAC Namehydrogen hydrazine chloride
Molecular FormulaClH5N2
InChI KeyBIVUUOPIAYRCAP-UHFFFAOYSA-N
SMILES[H+].[Cl-].NN
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
FormPowder or crystals or crystalline powder
Assay (unspecified)≥98.0%
Appearance (Color)White
Hydrazine dihydrochloride is used as chlorine scavenger in HCl gas streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only