Search
전체검색
View additional product information for Al-23 Solid Rod, Round;Diameter (mm), 5 - FAQs (032931.DT, 032931.DR, 032931.DW, 032931.QAT, 032931.EH)
1 product FAQs found
The maximum working temperature for the Al-23 Tube is 1950 degrees C.