Al-23 Solid Rod, Round, Dia.: 6mm, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Al-23 Solid Rod, Round, Dia.: 6mm, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
032933.DR
also known as 032933-DR
200 mm
Catalog number 032933.DR
also known as 032933-DR
Price (USD)
50.50
Each
Quantity:
200 mm
Request bulk or custom format
Price (USD)
50.50
Each
Specifications
Chemical Name or MaterialAl-23 Solid Rod
Recommended StorageAmbient temperatures
TSCAU
FormRod
Quantity200 mm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Frequently asked questions (FAQs)
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.