Al-23 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 15 x 10 x 8;Vol (ml), 0.7 - FAQs
View additional product information for Al-23 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 15 x 10 x 8;Vol (ml), 0.7 - FAQs (032950.KF)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.