Search
Search

| Catalog Number | Capacity (Metric) | Diameter (Metric) Outer | Diameter (Metric) Inner | Height (Metric) |
|---|---|---|---|---|
| 032953.KF also known as 032953-KF | 40 mL | 35mm | 30mm | 50mm |
| 032955.KF also known as 032955-KF | 110 mL | 50mm | 44mm | 75mm |
| 032954.KF also known as 032954-KF | 60 mL | 40mm | 36mm | 60mm |
| 032952.KF also known as 032952-KF | 20 mL | 30mm | 26mm | 40mm |
| 032951.KF also known as 032951-KF | 6 mL | 20mm | 16mm | 30mm |