Al-25 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 160 x 145 x 125;Vol (ml), 1760 - FAQs
Al-25 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 160 x 145 x 125;Vol (ml), 1760 - FAQs
View additional product information for Al-25 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 160 x 145 x 125;Vol (ml), 1760 - FAQs (032969.KF)
1 product FAQs found
What is the maximum working temperature of Al-25?
The maximum working temperature for Al-25 is 1950 degrees C.