Search
View additional product information for Al-25 Rectangular Plate;Length (mm), 115;Width (mm), 115;Thickness (mm), 15 - FAQs (033160.KF)
1 product FAQs found
The maximum working temperature for Al-25 is 1950 degrees C.