Al-23 Combustion Tray;L1 (mm), 75;L2 (mm), 73;B1 (mm), 50;B2 (mm), 48;H (mm), 20;W (mm), 4;Vol, 45.0 - FAQs
Al-23 Combustion Tray;L1 (mm), 75;L2 (mm), 73;B1 (mm), 50;B2 (mm), 48;H (mm), 20;W (mm), 4;Vol, 45.0 - FAQs
View additional product information for Al-23 Combustion Tray;L1 (mm), 75;L2 (mm), 73;B1 (mm), 50;B2 (mm), 48;H (mm), 20;W (mm), 4;Vol, 45.0 - FAQs (033185.KF)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.