Al-23 Grinding Pieces;Ht (mm), 10;Pcs/Kg, 440 - FAQs
Al-23 Grinding Pieces;Ht (mm), 10;Pcs/Kg, 440 - FAQs
View additional product information for Al-23 Grinding Pieces, H: 10mm, 440 Pieces/Kg, Thermo Scientific Chemicals - FAQs (033190.A1)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.