Thermo Scientific Chemicals

Oxalic acid dihydrate, ACS, 99.5-102.5%, Thermo Scientific Chemicals

Catalog number: 033262.A1
1 kg, Each
Thermo Scientific Chemicals

Oxalic acid dihydrate, ACS, 99.5-102.5%, Thermo Scientific Chemicals

Catalog number: 033262.A1
1 kg, Each
Quantity
Catalog number: 033262.A1
also known as 033262-A1
Price (USD)
Price: 135.00
Online price: 114.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6153-56-6
IUPAC Name
oxalic acid dihydrate
Molecular Formula
C2H6O6
InChI Key
GEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILES
O.O.OC(=O)C(O)=O
Residue on ignition
0.01%
Impurity content
Cl 0.002%
Impurity content
Nitrogen compounds (as N) 0.001%
Impurity content
Substances darkened by hot sulfuric acid P.T.
Impurity content
SO₄ 0.005%

Description

As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing and as a reagent for dehydration and condensation Oxalic acid dihydrate acts as an analytical reagent, as a reducing agent for photographic films and as mordant in the dyeing process. It is used as a plating agent, a surface treating agent, a grinding agent, a precipitating agent in rare-earth mineral processing, a bleaching agent in textile and a purifying agent in the pharmaceutical industry. It plays an important role in antibiotic medication like oxytetracycline and chloramphenicol. It finds application in water treatment plants to remove calcium waste water. It is also used as a buffer in chromatographic separation as well as bleach for wood, removing black stains caused by water penetration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing, and as a reagent for dehydration and condensation
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text