Thermo Scientific Chemicals

Iron(III) nitrate nonahydrate, ACS, 98.0-101.0%, Thermo Scientific Chemicals

Catalog number: 033315.F7
5 x 2 kg, Each
Thermo Scientific Chemicals

Iron(III) nitrate nonahydrate, ACS, 98.0-101.0%, Thermo Scientific Chemicals

Catalog number: 033315.F7
5 x 2 kg, Each
Quantity
Catalog number: 033315.F7
also known as 033315-F7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7782-61-8
IUPAC Name
iron(3+) nonahydrate trinitrate
Molecular Formula
FeH18N3O18
InChI Key
SZQUEWJRBJDHSM-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Impurity content
Ca 0.01%
Impurity content
Na 0.05%
Impurity content
Cl 5ppm
Impurity content
K 0.005%
Impurity content
Mg 0.005%

Description

It is used as catalyst for the synthesis of sodium amide from sodium and ammonia, and for etching of silver and silver alloys. Ferric nitrate is used in the synthesis of organically templated iron phosphates, and acts as an efficient catalyst in enamination of beta dicarbonyl compounds to beta enaminones. Ferric nitrate impregnated clays are useful as oxidizing agents. Clayfen, produced from ferric nitrate and montmorillonite, is used for the oxidation of thiols to disulfide, and alcohols to aldehydes. Ferric nitrate is useful for regio- and stereo-selective nitration of various aromatic, aliphatic, and heteroaromatic olefins. This reaction provides nitroolefins with excellent E-selectivity. With Keggin-type heteropoly acids, it selectively oxidizes various hydroxy groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text