Sodium hypochlorite, 11-15% available chlorine
Sodium hypochlorite, 11-15% available chlorine
Sodium hypochlorite, 11-15% available chlorine
Thermo Scientific Chemicals

Sodium hypochlorite, 11-15% available chlorine

CAS: 7681-52-9 | ClNaO | 74.439 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 g
2 kg
10 kg
產品號碼 033369.36
亦稱為 033369-36
價格 (TWD)
-
Quantity:
500 g
Request bulk or custom format
化學識別
CAS7681-52-9
IUPAC Namesodium hypochlorite
Molecular FormulaClNaO
InChI KeySUKJFIGYRHOWBL-UHFFFAOYSA-N
SMILES[Na+].[O-]Cl
檢視更多
規格Specification Sheet規格表
Appearance (Color)Pale yellow to green
FormLiquid
Assay (unspecified)10.5-15.5% available chlorine
Sodium hypochlorite is used as a disinfectant and a bleaching agent. It is used as a destainer involved in removing of mold stains, dental stains arises by fluorosis. It finds application in laundry detergents. Its aqueous solution is used as household bleach for sanitizing smooth surfaces. In a water treatment plant, it is utilized for treating dilute cyanide waste water like electroplating wastes. It is an oxidizing agent used with phase transfer catalyst to convert alcohols into corresponding carbonyl compounds. It is an important reactant in Hofmann rearrangement to prepare primary amine from primary amide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only