Thermo Scientific Chemicals

Sodium thiocyanate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 033388.A3
2 kg, Each
Thermo Scientific Chemicals

Sodium thiocyanate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 033388.A3
2 kg, Each
Quantity
Catalog number: 033388.A3
also known as 033388-A3
Price (USD)
Price: 312.00
Online price: 265.65
Your price:
Quantity
-

Chemical Identifiers

CAS
540-72-7
IUPAC Name
sodium cyanosulfanide
Molecular Formula
CNNaS
InChI Key
VGTPCRGMBIAPIM-UHFFFAOYSA-M
SMILES
[Na+].[S-]C#N
Assay (unspecified)
98.0% NaSCN min
Impurity content
Fe 2ppm
Impurity content
NH₄ 0.002%
Impurity content
Na₂CO₃ 0.2%
Impurity content
S 0.001%

Description

Sodium thiocyanate is used as a precursor for the synthesis of pharmaceuticals and specialty chemicals. It is used to prepare alkylthiocyanates by reacting with alkyl halides. It is involved nucleophilic substitution reactions with phase-transfer catalysts. It is a common laboratory reagent used to test presence of Fe3+ ions. It acts as a spinning solvent for polyacrylonitrile in fiber industry, photographic stabilizer, corrosion inhibitor, polymer stabilizer and hardening accelerator for cement and concrete mixes. Further, it is also used in both the crystallization and purification of proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text