Orthophosphoric acid, 33% (w/w), Biotech Reagent, GMP, J.T.Baker™
Avantor™

Orthophosphoric acid, 33% (w/w), Biotech Reagent, GMP, J.T.Baker™

This product is GMP Manufactured. Made from U.S.P. Purified Water and Phosphoric Acid, N.F. (Multicompendial) which meets B.P. and Ph.Eur. Chemical specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
19 L
200 L
Catalog number 0334-07
Price (USD)
-
Quantity:
19 L
This product is GMP Manufactured. Made from U.S.P. Purified Water and Phosphoric Acid, N.F. (Multicompendial) which meets B.P. and Ph.Eur. Chemical specifications.
This product is GMP Manufactured. Made from U.S.P. Purified Water and Phosphoric Acid, N.F. (Multicompendial) which meets B.P. and Ph.Eur. Chemical specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS7664-38-2
Chemical Name or MaterialOrthophosphoric acid
ColorColorless
Melting Point28°C
Molecular FormulaH3PO4
PackagingPoly bottle, corrugated shipper
SynonymPhosphoric acid
GradeBiotech
Molecular Weight (g/mol)98
Percent Purity0.33%
Quantity19 L
Unit Size19 L