Orthophosphoric acid, 33% (w/w), Biotech Reagent, GMP, J.T.Baker™
Avantor™

Orthophosphoric acid, 33% (w/w), Biotech Reagent, GMP, J.T.Baker™

This product is GMP Manufactured. Made from U.S.P. Purified Water and Phosphoric Acid, N.F. (Multicompendial) which meets B.P. and Ph.Eur.자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
수량:
19 L
200 L
카탈로그 번호 0334-09
제품 가격(KRW)
-
수량:
200 L
This product is GMP Manufactured. Made from U.S.P. Purified Water and Phosphoric Acid, N.F. (Multicompendial) which meets B.P. and Ph.Eur. Chemical specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
사양
CAS7664-38-2
화학물질 이름 또는 재질Orthophosphoric acid
색상Colorless
용융점28°C
분자식H3PO4
포장Polyethylene drum
동의어Phosphoric acid
등급Biotech
분자량(g/mol)98
백분율 순도0.33%
수량200 L
Unit Size200 L