Dionex™ Chelation Chromatography Reagents
Actual product may vary
Thermo Scientific™

Dionex™ Chelation Chromatography Reagents

Facilitate trace and ultra trace determination of transition and lanthanide metals with this range of dedicated Thermo Scientific™ Dionex™ Chelation Chromatography Reagents.
Have Questions?
Catalog NumberDescriptionQuantity
0334402.0 M Ammonium Acetate1 L
Catalog number 033440
Price (TWD)
-
Description:
2.0 M Ammonium Acetate
Quantity:
1 L
Designed to facilitate trace and ultra trace determination of transition and lanthanide metals, Thermo Scientific™ Dionex™ Chelation Chromatography Reagents are dedicated for use with the Dionex IC/ICP Interface Module for ultimate ease of use. Available in 1L quantities, reagents include 2.0M nitric acid, 2.0M ammonium acetate, and 0.1M ammonium nitrate. Order each in single or six-pack order quantities. Only high-purity, reagent-grade chemicals and water (18MO-cm), quality-control checked for ionic contamination, are used in Dionex reagents, standards and eluent concentrates.
  • Only high-purity, reagent-grade chemicals and water (18 MΩ-cm), quality-control checked for ionic contamination, are used
  • All standards are NIST™ traceable

Includes: A Certificate of Analysis is provided with each product.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceNIST Traceable
Concentration2.0 M
Description2.0 M Ammonium Acetate
For Use With (Equipment)Dionex Chelation Concentration System
HazardsIrritant
IncludesCertificate of Analysis
Quantity1 L
Standard ComponentsAmmonium Acetate Aqueous Solution
Standard Concentration2 M
StorageRoom Temperature
TypeChelation Chromatography Reagent
Unit SizeEach 1