Thermo Scientific Chemicals
Molecular sieves, 13X, 3-4mm (0.12-0.16in) dia. pellets, Thermo Scientific Chemicals
Catalog number: 033550.A7
5 kg, Each
Thermo Scientific Chemicals
Molecular sieves, 13X, 3-4mm (0.12-0.16in) dia. pellets, Thermo Scientific Chemicals
Catalog number: 033550.A7
5 kg, Each
Quantity
Catalog number: 033550.A7
also known as 033550-A7
Price (USD)
Price: 302.00
Online price: 256.65
Your price:
Quantity
-
Description
Some of the major applications include commercial gas drying, air plantfeed purification (simultaneous H2O and CO2 removal) and liquid hydrocarbon/natural gas sweetening (H2S and mercaptan removal).