Hydrochloric acid, 2.0 N, Biotech Reagent, GMP, J.T.Baker™
Avantor™

Hydrochloric acid, 2.0 N, Biotech Reagent, GMP, J.T.Baker™

Made from USP purified water and NF (multicompendial) hydrochloric acid. Meets BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.Read more
Have Questions?
Catalog number 0336-03
Price (EUR)
-
Made from USP purified water and NF (multicompendial) hydrochloric acid. Meets BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point100°C to 103°C
CAS7647-01-0
Chemical Name or MaterialHydrochloric acid
ColorColorless
Molecular FormulaHCl
PackagingPoly bottle, pressure bottom
SynonymHydrochloric acid, Hydrogen chloride solution
GradeBiotech
Molecular Weight (g/mol)36.46
Quantity4 L
Unit SizeEach