Sodium hydroxide, 50% in aqueous solution, Biotech Reagent, GMP, J.T.Baker™
Avantor™

Sodium hydroxide, 50% in aqueous solution, Biotech Reagent, GMP, J.T.Baker™

Made from USP purified water and sodium hydroxide pellets, NF (Multi-Compendial). Meets BP, Ph.Eur. and JP chemical specifications.Read more
Have Questions?
Change viewbuttonViewtableView
Quantity:
19 L
200 L
Packaging:
View all
Polyethylene drum
Catalog number 0339-09
Price (KRW)
-
Quantity:
200 L
Packaging:
Polyethylene drum
Made from USP purified water and sodium hydroxide pellets, NF (Multi-Compendial). Meets BP, Ph.Eur. and JP chemical specifications.
Specifications
Boiling Point119°C to 135°C (1013 hPa)
CAS1310-73-2
Chemical Name or MaterialSodium hydroxide
ColorColorless
Melting Point9°C to 10°C
Molecular FormulaNaOH
PackagingPolyethylene drum
SynonymSodium hydrate, Soda lye, Caustic soda, Lye
Molecular Weight (g/mol)40
Quantity200 L
Unit SizeEach