Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Fisher Scientific™

Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Have Questions?
카탈로그 번호웰 수두께(미터법)부피(미터법) 웰
03500163
FIS-03500163으로도 사용됨
100.075 cm9.8μL/ mm
카탈로그 번호 03500163
FIS-03500163으로도 사용됨
제품 가격(KRW)
-
웰 수:
10
두께(미터법):
0.075 cm
부피(미터법) 웰:
9.8μL/ mm
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
  • Lightweight
  • Virtually unbreakable
사양
깊이(영국식 단위)0.51 in.
깊이(미터법)1.3 cm
물질PTFE
웰 수10
두께(영국식 단위)0.03 in.
두께(미터법)0.075 cm
부피(미터법) 웰9.8μL/ mm
폭(미터법)0.48 cm
용도 (장비)SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
유형Comb
Unit SizeEach