Dionex™ Ion Pairing Reagents
Actual product may vary
Dionex™ Ion Pairing Reagents
Dionex™ Ion Pairing Reagents
Dionex™ Ion Pairing Reagents
Dionex™ Ion Pairing Reagents
Thermo Scientific™

Dionex™ Ion Pairing Reagents

Highly purified ion-pairing reagents are used in mobile phase ion chromatography (MPIC).

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
035360Tetrabutylammonium Hydroxide
035362Octanesulfonic Acid
Catalog number 035360
Price (USD)
381.00
Each
Description:
Tetrabutylammonium Hydroxide
Price (USD)
381.00
Each

Highly purified Thermo Scientific™ Dionex™ Ion Pairing Reagents are used in mobile phase ion chromatography (MPIC), combining reversed-phase ion-pair chromatography with chemical suppression.

  • Prepared by the dissolution of high-purity chemicals in >18.2MΩ-cm deionized water, which was tested by ion chromatography for ionic contaminants
  • Bottle label states the nominal concentration value for informational purposes only - actual concentration is within ±2.5% and was determined by one or more of the following proven reliable methods of analysis: ion chromatography, UV spectrophotometry (using NIST or USP Reference Material where applicable), titration (using NIST™ Reference Material), and gravimetric preparation
  • Concentration: 0.1M
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionTetrabutylammonium Hydroxide
For Use With (Application)MPIC
HazardsCorrosive, Irritant
OperationUsed in mobile phase ion chromatography (MPIC), combining reversed-phase ion-pair chromatography with chemical suppression.
QuantityEach
Standard ComponentsTetrabutylammonium Hydroxide in Deionized Water
Standard Concentration0.1 M
StorageRoom Temperature
Volume (Metric)500 mL
TypeIon Pairing Reagent
Unit SizeEach