This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035417.BG
also known as 035417-BG
Price (EUR)
-
Quantity:
2 x 100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialInstrument Calibration(ICAL) Standards Kit
MDL NumberMFCD00147264
Physical FormLiquid
Quantity2 x 100 mL
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.