Especificaciones
Nombre del producto químico o materialContinuing Calibration Verification (CCV) Standards Kit
Concentration or Composition (by Analyte or Components)(CCV) solución A, (CCV) solución B
Densidad1g/cm3 at 20°C
Peligro para la salud 1H314:Provoca quemaduras graves en la piel y lesiones oculares.
Peligro para la salud 2Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Ver más