Thermo Scientific Chemicals

Acetic acid, 4% v/v aq. soln., Thermo Scientific Chemicals

500 mL, Each
Thermo Scientific Chemicals

Acetic acid, 4% v/v aq. soln., Thermo Scientific Chemicals

500 mL, Each
Quantity
Catalog number: 035572.AP
also known as 035572-AP
Price (USD)
Price: 26.60
Your Price:
Quantity
-
Assay (Titration)
4.0 % +/- 0.2 % ( v / v)
Form
clear liquid
Appearance (Color)
Colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only