Thermo Scientific Chemicals

Potassium hydrogen phthalate, 0.05N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035603.K2
1 L, Each
Thermo Scientific Chemicals

Potassium hydrogen phthalate, 0.05N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035603.K2
1 L, Each
Quantity
Catalog number: 035603.K2
also known as 035603-K2
Price (TWD)

Chemical Identifiers

CAS
877-24-7
IUPAC Name
potassium 2-carboxybenzoate
Molecular Formula
C8H5KO4
InChI Key
IWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES
[K+].OC(=O)C1=CC=CC=C1C([O-])=O
Assay from Supplier's CofA
0.0500N ? 0.0001N
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text