Thermo Scientific Chemicals

Nitric acid, 1.0N Standardized Solution, Thermo Scientific Chemicals

500 mL, Each
Thermo Scientific Chemicals

Nitric acid, 1.0N Standardized Solution, Thermo Scientific Chemicals

500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 035624.AP
also known as 035624-AP
Price (USD)
Price: 42.40
Your Price:
Quantity
-

Chemical Identifiers

CAS
7697-37-2
IUPAC Name
nitric acid
Molecular Formula
HNO3
InChI Key
GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES
O[N+]([O-])=O
Concentration
1.0N

Description

Nitric acid, 1.0N Standardized Solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only