Sodium hydroxide, 0.5N Standardized Solution
Sodium hydroxide, 0.5N Standardized Solution
Sodium hydroxide, 0.5N Standardized Solution
Thermo Scientific Chemicals

Sodium hydroxide, 0.5N Standardized Solution

Sodium hydroxide, HNaO, CAS Number-1310-73-2, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, Colorless solution, CHEBI:32145, DOT Class: 8, Packaging Group: III, 215-185-5, 40 | CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
20 L
Catalog number 035627.K2
also known as 035627-K2
Price (EUR)
35,20
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
35,20
Each
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Concentration0.500N ± 0.14%
Sodium hydroxide, 0.5N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only