Iodine, 0.1N Standardized Solution
Iodine, 0.1N Standardized Solution
Iodine, 0.1N Standardized Solution
Thermo Scientific Chemicals

Iodine, 0.1N Standardized Solution

CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 L
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 035634.K7
價格 (TWD)
-
Quantity:
4 L
Request bulk or custom format
化學識別
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
檢視更多
規格Specification Sheet規格表
Appearance (Color)Dark brown
FormLiquid
Concentration0.100N ± 0.19%
Iodine, 0.1N standardized solution is used for accurate volumetric analysis. It is also used as a disinfectant and antibacterial agent in solutions, tinctures and salves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only