Informational:Try chem dex – the new web app for synthetic chemists
Thermo Scientific Chemicals

Sodium hydroxide, 20% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 035637.K7
4 L, Each
Thermo Scientific Chemicals

Sodium hydroxide, 20% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 035637.K7
4 L, Each
Quantity
Catalog number: 035637.K7
also known as 035637-K7
Price (USD)
Price: 83.40
Your Price:
Quantity
-

Chemical Identifiers

CAS
1310-73-2
IUPAC Name
sodium hydroxide
Molecular Formula
HNaO
InChI Key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES
[OH-].[Na+]
Assay (Aqueous acid-base Titration)
20.0% Sodium hydroxide

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text