Boron silicide, 98% (metals basis)
Boron silicide, 98% (metals basis)
Boron silicide, 98% (metals basis)
Thermo Scientific Chemicals

Boron silicide, 98% (metals basis)

CAS: 12008-29-6 | B6Si | 92.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 035670.09
also known as 035670-09
Price (USD)
108.65
Special offer
Online exclusive
Ends: 30-Jun-2026
128.00
Save 19.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
108.65
Special offer
Online exclusive
Ends: 30-Jun-2026
128.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS12008-29-6
IUPAC Name1-sila-2,3,4,5,6,7-hexaborapentacyclo[3.2.0.0¹,⁴.0²,⁷.0³,⁶]heptane
Molecular FormulaB6Si
InChI KeyZRBFEDMQRDRUDG-UHFFFAOYSA-N
SMILESB12B3B4B5B(B14)[Si]235
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities2% max.
Boron silicide is used in chemical research, pharmaceuticals and as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only