Cadmium bromide, ultra dry, 99.999% (metals basis)
Cadmium bromide, ultra dry, 99.999% (metals basis)
Cadmium bromide, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Cadmium bromide, ultra dry, 99.999% (metals basis)

CAS: 7789-42-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035682.06
also known as 035682-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7789-42-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.001% max.
Particle Size-10 Mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only