Chromium(III) chloride, anhydrous, 99.9% (metals basis)
Thermo Scientific Chemicals

Chromium(III) chloride, anhydrous, 99.9% (metals basis)

CAS: 10025-73-7 | CrCl3
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 035691.06
also known as 035691-06
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.40
Save 6.75 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.40
Save 6.75 (15%)
Each
Chemical Identifiers
CAS10025-73-7
IUPAC Namechromium(3+) hexahydrate trichloride
Molecular FormulaCl3CrH12O6
InChI KeyLJAOOBNHPFKCDR-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. It is used as a catalyst, for organic synthesis, for manufacturing dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only