Manganese(II) iodide, ultra dry, 99.99% (metals basis)
Manganese(II) iodide, ultra dry, 99.99% (metals basis)
Manganese(II) iodide, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Manganese(II) iodide, ultra dry, 99.99% (metals basis)

Often used in the lighting industry and it is a source of its derivatives. | CAS: 7790-33-2 | I2Mn
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Packaging:
View all
Ampouled under argon
Catalog number 035710.04
also known as 035710-04
Price (USD)
146.65
Special offer
Online exclusive
Ends: 30-Jun-2026
172.00
Save 25.35 (15%)
Each
Quantity:
2 g
Packaging:
Ampouled under argon
Request bulk or custom format
Price (USD)
146.65
Special offer
Online exclusive
Ends: 30-Jun-2026
172.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS7790-33-2
IUPAC Namemanganese(2+) diiodide
Molecular FormulaI2Mn
InChI KeyQWYFOIJABGVEFP-UHFFFAOYSA-L
SMILES[Mn++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water< 100 ppm
Appearance (Color)Pink
Total Metal Impurities0.01% max.
FormPowder/flakes
It is often used in the lighting industry and it is a source of its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only