Thermo Scientific Chemicals

Sodium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 035716.22
100 g, Each
Thermo Scientific Chemicals

Sodium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 035716.22
100 g, Each
Quantity
Catalog number: 035716.22
Price (USD)
Price: 462.00
Online price: 392.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7647-14-5
IUPAC Name
sodium chloride
Molecular Formula
ClNa
InChI Key
FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES
[Na+].[Cl-]
Appearance (Color)
White
Form
Beads
Residual water
< 100 ppm
Total Metal Impurities
0.01% max.
Particle Size
-10 Mesh

Description

Sodium chloride is widely used as a food additive, and a preservative. It is employed in the Chloralkali process for producing chlorine and sodium hydroxide, in the Solvay process for producing sodium carbonate and calcium chloride, in the Mannheim process and in the Hargreaves process for producing sodium sulfate and hydrochloric acid. It is used in the manufacture of sodium chlorate, water softening and de-icing of roadways. Further it is used in several industries like pharmaceutical, textile, dyeing, rubber food and agriculture. It is used in the processing of steel, copper, vanadium, and other metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text