Thermo Scientific Chemicals

Strontium chloride, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 035719.06
5 g, Each
Thermo Scientific Chemicals

Strontium chloride, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 035719.06
5 g, Each
Quantity
Catalog number: 035719.06
also known as 035719-06
Price (USD)
Price: 98.80
Online price: 84.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10476-85-4
IUPAC Name
strontium(2+) hexahydrate dichloride
Molecular Formula
Cl2H12O6Sr
InChI Key
AMGRXJSJSONEEG-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
Appearance (Color)
White
Form
Beads
Residual water
< 100 ppm
Total Metal Impurities
0.005% max.
Particle Size
-10 Mesh

Description

Strontium chloride  is used as a source of redness in fireworks. Strontium chloride is often used as a red coloring agent in pyrotechnics. It is employed in small quantities in glass-making and metallurgy. The radioactive isotope strontium-89, used for the treatment of bone cancer, is usually administered in the form of strontium chloride. Sea water aquaria require small amounts of strontium chloride, which is consumed in the production of the exoskeletons of certain plankton. SrCl2 is useful in reducing tooth sensitivity by forming a barrier over microscopic tubules in the dentin containing nerve endings that have become exposed by gum recession. strontium chloride exposure induces parthenogenetic activation of oocytes[6] which is used in developmental biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text