Titanium, plasma standard solution, Specpure™ Ti 10,000μg/mL
Titanium, plasma standard solution, Specpure™ Ti 10,000μg/mL
Titanium, plasma standard solution, Specpure™ Ti 10,000μg/mL
Thermo Scientific Chemicals

Titanium, plasma standard solution, Specpure™ Ti 10,000μg/mL

Have Questions?
變更視圖buttonViewtableView
Quantity:
50 mL
100 mL
500 mL
產品號碼 035759.AD
亦稱為 035759-AD
價格 (TWD)
-
Quantity:
50 mL
Request bulk or custom format
規格
Chemical Name or MaterialTitanium plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only