Lutetium(III) chloride, ultra dry, 99.98% (REO)
Lutetium(III) chloride, ultra dry, 99.98% (REO)
Lutetium(III) chloride, ultra dry, 99.98% (REO)
Thermo Scientific Chemicals

Lutetium(III) chloride, ultra dry, 99.98% (REO)

CAS: 10099-66-8
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 035802.03
亦稱為 035802-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS10099-66-8
IUPAC Namelutetium(3+) trichloride
Molecular FormulaCl3Lu
InChI KeyAEDROEGYZIARPU-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Lu+3]
檢視更多
規格Specification Sheet規格表
Residual water< 100 ppm
Total Rare Earth Oxide Impurities0.02% max.
Appearance (Color)White
FormPowder
It is used as an additive for chemical industry, single crystal scintillators, carbon arc rods, PET scan detectors, high refractive index glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only