Bis(cyclopentadienyl)hafnium dichloride
Bis(cyclopentadienyl)hafnium dichloride
Bis(cyclopentadienyl)hafnium dichloride
Thermo Scientific Chemicals

Bis(cyclopentadienyl)hafnium dichloride

CAS: 12116-66-4 | C10H10Cl2Hf | 379.58 g/mol
製品番号(カタログ番号) 036105.03
または、製品番号036105-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS12116-66-4
化学物質名または材質Bis(cyclopentadienyl)hafnium dichloride
DOT情報Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECS番号235-177-5
式量379.59
さらに表示
Starting material to prepare many kinds of hafnocene(II or IV) derivatives; in combination with AgI salt, is an effective activator for glycosyl fluoride. Pharmaceutical intermediates and alos used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only