Iridium foil, 0.5mm (0.02in) thick, typically 99.7+% (metals basis)
Iridium foil, 0.5mm (0.02in) thick, typically 99.7+% (metals basis)
Iridium foil, 0.5mm (0.02in) thick, typically 99.7+% (metals basis)
Iridium foil, 0.5mm (0.02in) thick, typically 99.7+% (metals basis)
Thermo Scientific Chemicals

Iridium foil, 0.5mm (0.02in) thick, typically 99.7+% (metals basis)

CAS: 7439-88-5 | Ir | 192.22 g/mol
Have Questions?
產品號碼Quantity
036149.FF25 x 25 mm
產品號碼 036149.FF
價格 (TWD)
-
申請報價
Quantity:
25 x 25 mm
Request bulk or custom format
規格
Assay Percent Notes(metals basis)
CAS7439-88-5
ChEBICHEBI:49666
Chemical Name or MaterialIridium foil
EINECS Number231-095-9
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only