Titanium(IV) oxide, anatase, 99.6% (metals basis)
Titanium(IV) oxide, anatase, 99.6% (metals basis)
Titanium(IV) oxide, anatase, 99.6% (metals basis)
Thermo Scientific Chemicals

Titanium(IV) oxide, anatase, 99.6% (metals basis)

CAS: 1317-70-0 | O2Ti | 79.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 036199.14
also known as 036199-14
Price (USD)
45.30
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
45.30
Each
Chemical Identifiers
CAS1317-70-0
IUPAC Namedioxotitanium
Molecular FormulaO2Ti
InChI KeyGWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILESO=[Ti]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.4% max.
Titanium(IV) oxide is used as UV-resistant cloth, skin protecting cream, face washing milk, cream milk, powder make-up. It is also used as photocatalyst, self-cleaning glass, self-cleaning ceramics, antibacterial material, air purification, sewage handling, chemical industry. It is widely utilized in cosmetics, sunscreen cream, natural white moisture protection cream, beauty and whitening cream, dawn and night cream, moistening refresher, vanishing cream, chemical fiber, plastics, printing ink coating. It is likewise utilized for improving the impressionability and opacity of the paper and used for producing titanium, ferrotitanium alloy, carbide alloy etc. in the metallurgical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the difference between Titanium (IV) oxide, rutile and Titanium (IV) oxide, anatase?

Rutile and anatase are polymorphs of TiO2, which are both naturally occurring. Rutile is a source for titanium smelting and is used in the semiconductor industry to produce dye-based solar cells (Grätzel cells). Anatase shows a high photocatalytic activity, which limits the usability as a pigment in polymers, but also opens certain applications as a catalyst. Anatase has a higher photocatalytic activity than rutile but is thermodynamically less stable.