Lithium carbonate, ACS, 99.0% min
Lithium carbonate, ACS, 99.0% min
Lithium carbonate, ACS, 99.0% min
Lithium carbonate, ACS, 99.0% min
Lithium carbonate, ACS, 99.0% min
Thermo Scientific Chemicals

Lithium carbonate, ACS, 99.0% min

Maximum level of impurities: Insoluble in dilute hydrochloric acid 0.01%, Cl 0.005%, NO3 5ppm, Sulfur compounds (as SO4) 0.2%, NH4 5ppm, Heavy Metals (as Pb) 0.002%, Fe 0.002%, Ca 0.01%, K 0.01%, Na 0.1% | CAS: 554-13-2 | CLi2O3 | 73.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 036225.A3
also known as 036225-A3
Price (EUR)
906,00
Each
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (EUR)
906,00
Each
Add to cart
Chemical Identifiers
CAS554-13-2
IUPAC Namedilithium(1+) carbonate
Molecular FormulaCLi2O3
InChI KeyXGZVUEUWXADBQD-UHFFFAOYSA-L
SMILES[Li+].[Li+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder
Assay (unspecified)≥99.0% Li2CO3
Chloride content≤0.005%
Impurity contentCalcium (Ca): ≤0.01%
View more

Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only