Aluminon, ACS
Aluminon, ACS
Aluminon, ACS
Thermo Scientific Chemicals

Aluminon, ACS

CAS: 569-58-4 | C22H23N3O9 | 473.438 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 036239.14
also known as 036239-14
Price (USD)
53.65
Online exclusive
59.80
Save 6.15
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
53.65
Online exclusive
59.80
Save 6.15
Each
Chemical Identifiers
CAS569-58-4
IUPAC Nametriammonium 5-[(3-carboxylato-4-hydroxyphenyl)(3-carboxylato-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoate
Molecular FormulaC22H25N3O9
InChI KeyUDPQMNAHKKTHHI-UHFFFAOYNA-N
SMILES[NH4+].[NH4+].[NH4+].OC1=CC=C(C=C1C([O-])=O)C(C1C=CC(=O)C(=C1)C([O-])=O)C1=CC=C(O)C(=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSensitivity to aluminum P.T.
Residue on ignition0.2%
Insoluble material0.1%
It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types. Its neuroprotective effect, perhaps due to its ability to prevent down-regulation of Ca2+ impermeable GluR2 receptors or to its ability to inhibit calpain, a Ca2+-activated protease that is activated during apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the solubility of Aluminon?

Aluminon is soluble in water at a concentration of ~800 g/L at 20°C. It is slightly soluble in ethanol and insoluble in petroleum ether.