Thermo Scientific Chemicals

Aluminón, ACS, Thermo Scientific Chemicals

Número de catálogo: 036239.22
100 g, Each
Thermo Scientific Chemicals

Aluminón, ACS, Thermo Scientific Chemicals

Número de catálogo: 036239.22
100 g, Each
Cantidad
Número de catálogo: 036239.22
también denominado 036239-22
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
569-58-4
IUPAC Name
triammonium 5-[(3-carboxylato-4-hydroxyphenyl)(3-carboxylato-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoate
Molecular Formula
C22H25N3O9
InChI Key
UDPQMNAHKKTHHI-UHFFFAOYNA-N
SMILES
[NH4+].[NH4+].[NH4+].OC1=CC=C(C=C1C([O-])=O)C(C1C=CC(=O)C(=C1)C([O-])=O)C1=CC=C(O)C(=C1)C([O-])=O
Insoluble material
0.1%
Comment
Sensitivity to aluminum P.T.
Residue on ignition
0.2%

Descripción

It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types. Its neuroprotective effect, perhaps due to its ability to prevent down-regulation of Ca2+ impermeable GluR2 receptors or to its ability to inhibit calpain, a Ca2+-activated protease that is activated during apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing