Thermo Scientific Chemicals

Mercury(II) oxide, yellow, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036261.14
25 g, Each
Thermo Scientific Chemicals

Mercury(II) oxide, yellow, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036261.14
25 g, Each
Quantity
Catalog number: 036261.14
also known as 036261-14
Price (USD)

Chemical Identifiers

CAS
21908-53-2
IUPAC Name
oxomercury
Molecular Formula
HgO
InChI Key
UKWHYYKOEPRTIC-UHFFFAOYSA-N
SMILES
O=[Hg]
Assay (unspecified)
99.0% HgO min
Comment
Residue after reduction 0.05%
Impurity content
Cl 0.025%
Impurity content
Fe 0.003%
Impurity content
Insoluble in dilute hydrochloric acid 0.03%

Description

Mercury(II) oxide production of mercury. It is also used as a material for cathodes for mercury batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text