Praseodymium(III, IV) oxide, 99.9% (REO)
Praseodymium(III, IV) oxide, 99.9% (REO)
Praseodymium(III, IV) oxide, 99.9% (REO)
Thermo Scientific Chemicals

Praseodymium(III, IV) oxide, 99.9% (REO)

CAS: 12037-29-5 | O11Pr6 | 1021.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 036276.18
also known as 036276-18
Price (EUR)
222,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
222,00
Each
Chemical Identifiers
CAS12037-29-5
IUPAC Namehexapraseodymium(3+) undecaoxidandiide
Molecular FormulaO11Pr6
InChI KeyHPZIIFVSYNLWNX-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Praseodymium(III, IV) oxide is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only