Acetic acid, glacial, ACS, 99.7+%
Acetic acid, glacial, ACS, 99.7+%
Acetic acid, glacial, ACS, 99.7+%
Thermo Scientific Chemicals

Acetic acid, glacial, ACS, 99.7+%

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2 L
4 x 500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036289.K3
also known as 036289-K3
Price (EUR)
100,00
Each
Quantity:
2 L
Request bulk or custom format
Price (EUR)
100,00
Each
Chemical Identifiers
CAS64-19-7
IUPAC Nameacetic acid
Molecular FormulaC2H4O2
InChI KeyQTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILESCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥99.7%
CommentDilution Test P.T.
Free base (titration)0.0004meq/g Max
Impurity content(CH₃CO)₂O 0.01% Max
View more
It is used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as table vinegar, as preservative and as intermediate in the chemical industry. It is useful for the extraction of glazed ceramic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only